missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Coverall (all)

Catalog No. 19162536
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
X-Large
Color:
Blue
White
Catalog No. Size Color
19162536 Large Blue
19162535 Medium Blue
19162537 X-Large Blue
19162538 2X-Large Blue
19162539 3X-Large Blue
19162541 5X-Large Blue
19162528 Medium White
19162529 Large White
19162530 X-Large White
19162531 2X-Large White
19162532 3X-Large White
19162533 4X-Large White
12 options

Catalog No. 19162536

Supplier: DuPont™ PB125SBULG002500

Only null left

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model PB125S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less