missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Coverall (all)

Catalog No. 19162535
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Color:
Blue
White
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19162535 Medium Blue
19162536 Large Blue
19162537 X-Large Blue
19162538 2X-Large Blue
19162539 3X-Large Blue
19162541 5X-Large Blue
19162528 Medium White
19162529 Large White
19162530 X-Large White
19162531 2X-Large White
19162532 3X-Large White
19162533 4X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19162535 Supplier DuPont™ Supplier No. PB125SBUMD002500

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model PB125S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less