missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Hooded Coverall

Catalog No. 19162527
Change view
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19162527 Blue 5X-Large
19162514 White Medium
19162515 White Large
19162519 White 4X-Large
19162518 White 3X-Large
19162522 Blue Large
19162517 White 2X-Large
19162521 Blue Medium
19162516 White X-Large
19162520 White 5X-Large
19162523 Blue X-Large
19162526 Blue 4X-Large
19162524 Blue 2X-Large
19162525 Blue 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19162527 Supplier DuPont™ Supplier No. PB122SBU5X002500

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Attached skid-resistant boots
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Coveralls
Size 5X-Large
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ASTM D1117
Model PB122S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less