Learn More
DuPont™ ProShield™ 10 Hooded Coverall
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB122SWH4X002500
 View more versions of this product
Catalog No. 19162519
Only null left
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Attached skid-resistant boots
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Attached
Serged
ASTM D1117
General Maintenance, Janitorial, Cleaning
4X-Large
ProShield
Single-use
Attached Boots
Hooded
Front Zipper
Non-sterile
PB122S
25/Cs.
Product Suggestions
Product Suggestions