missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProClean™ Frock

Catalog No. 19088388
Change view
Click to view available options
Closure Type:
Snaps
Zipper
Size:
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Size
19088388 Zipper 3X-Large
190881019 Zipper Large
190881018 Zipper X-Large
190881029 Snaps X-Large
19088387 Zipper 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19088388 Supplier DuPont™ Supplier No. PC271SWH3X00300B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Bulk packaged in double transparent poly liners.

Specifications

Size 3X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Cleanroom Class Class 1000 (ISO 6)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) General Maintenance, Janitorial, Cleaning, Light Industrial Applications
Show More Show Less