Learn More
DuPont™ ProClean™ Frock
Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.
Supplier: DuPont™ PC271SWH2X00300B
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding
- Frocks provide body-length jacket design with front snap closures for easy donning and doffing
- Bulk packaged in double transparent poly liners.
Specifications
| Controlled Environments Frock | |
| Microporous Composite Fabric | |
| Non-sterile | |
| Elastic | |
| Class 1000 (ISO 6) |
| 2X-Large | |
| White | |
| Zipper | |
| Serged |