missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 60 Hooded Coverall

Catalog No. 29634246
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29634246 Small
29634248 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29634246 Supplier DuPont™ Supplier No. NG127SWHSM0025NP

Coveralls with respirator fit hood are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Latex Free Yes
Model NG127S
Size (English) Chest 29.75 to 33.25 in.
Size (English) Inseam 26 in.
Size (Metric) Chest 75.56 to 84.45 cm
Size (Metric) Inseam 66.04 cm
Thickness 10 mil
For Use With (Application) Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
Length (English) Sleeve 30.75 in.
Length (Metric) Sleeve 78 cm
Height (English) 5 to 5.7 ft.
Height (Metric) 1.52 to 1.73 m
Quantity 25/Cs.
Show More Show Less