missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 60 Lab Coat

Catalog No. 29634925
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29634925 3X-Large
29634924 2X-Large
29634921 Medium
29634923 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29634925 Supplier DuPont™ Supplier No. NG212SWH3X003000

Limited-use general purpose lab coats are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.

  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material ProShield
Color White
Closure Type Snaps
Cuff Style Open
Disposable Limited-use
Sterility Non-sterile
Resistance Light Liquid Splash
No. of Pockets 2
Collar Type Lay Down
Model NG212S
Seam Type Serged
For Use With (Application) Automotive Refinishing, Waste Cleanup, Remediation
Show More Show Less