missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 60 Lab Coat
Limited-use general purpose lab coats are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.
Supplier: DuPont™ NG212SWHXL003000
Description
- Labcoat offers front snap closures for easy donning and doffing
- Provides enhanced frontal protection
- Laydown collar
- Open wrists
- Two front pockets for holding writing instruments, small tools or parts
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| General Purpose Lab Coat | |
| Unisex | |
| White | |
| Open | |
| Non-sterile | |
| 2 | |
| NG212S | |
| 39.75 to 43.25 in. | |
| 34.5 in. |
| X-Large | |
| ProShield | |
| Snaps | |
| Limited-use | |
| Light Liquid Splash | |
| Lay Down | |
| Serged | |
| Automotive Refinishing, Waste Cleanup, Remediation | |
| 87.63 cm |