missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 60 Lab Coat

Catalog No. 29634921
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29634921 Medium
29634924 2X-Large
29634925 3X-Large
29634923 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29634921 Supplier DuPont™ Supplier No. NG212SWHMD003000
Only null left

Limited-use general purpose lab coats are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.

  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material ProShield
Color White
Closure Type Snaps
Cuff Style Open
Disposable Limited-use
Sterility Non-sterile
Resistance Light Liquid Splash
No. of Pockets 2
Collar Type Lay Down
Model NG212S
Seam Type Serged
Size (English) Chest 33.25 to 36.75 in.
For Use With (Application) Automotive Refinishing, Waste Cleanup, Remediation
Length (English) Sleeve 32 in.
Length (Metric) Sleeve 81.28 cm
Show More Show Less