Learn More
DuPont™ IsoClean™ Eyes Only Cleanroom Hoods
GREENER_CHOICE
Catalog No. p-22256
Change view
Click to view available options
Size:
Medium
Large
Sterility:
Non-sterile
Sterile
Hood Type:
Clean and Sterile
Clean-processed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility Hood Type
190881134B Medium Non-sterile Clean-processed
190881134C Large Non-sterile Clean-processed
190881135B Medium Sterile Clean and Sterile
190881135C Large Sterile Clean and Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 190881134B Supplier DuPont™ Supplier No. IC666BWHMD01000C
Only null left
Tyvek™ cleanroom hood features bound seams for strength and durability and meets ISO Class 4 standards. Available in clean or clean and sterile versions.
  • Clean-processed garments offer low level of particle shedding
  • Bound seams are covered with garment fabric for reinforcement
  • Bound hood opening for lower particle shedding
  • Eyes-only opening
  • Snaps for adjustable fit
  • Available in medium or large sizes
  • Available in clean or clean and sterile versions
Specifications
Product Type Eyes Only
Size Medium
Material Tyvek
Color White
Closure Type Snap
Disposable Single-use
Model I666B
Sterility Non-sterile
Quantity 100/Cs.
Hood Type Clean-processed
Cleanroom Class Class 1-10 (ISO 3-4)
Show More Show Less