missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)

Catalog No. 296360976
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296360976 Medium
296360975 Small
190881121C Large
190881121D X-Large
190881121E 2X-Large
190881121F 3X-Large
190881121G 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 296360976 Supplier DuPont™ Supplier No. IC182BWHMD00250C

Tyvek IsoClean delivers an ideal balance of protection, durability, and comfort. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.

  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Raglan sleeve design for greater range of motion and comfort.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Size (English) Chest 36 to 38 in.
Size (English) Inseam 32.5 in.
Size (Metric) Chest 91.44 to 96.52 cm
Size (Metric) Inseam 82.55 cm
Thickness 5.4 mil
For Use With (Application) Premium protection for controlled environments and inherent barrier to dry particles, microorganisms, and non-hazardous liquids
Length (English) Sleeve 37 in.
Length (Metric) Sleeve 93.98 cm
Height (English) 5.5 to 5.10 ft.
Height (Metric) 167.64 to 155.44 cm
Packaging Clean-processed
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less