missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Tyvek IsoClean delivers an ideal balance of protection, durability, and comfort. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Supplier: DuPont™ IC182BWH3X00250C
Description
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle.
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing.
- Raglan sleeve design for greater range of motion and comfort.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Covered Elastic | |
| Zipper | |
| Non-sterile | |
| 35 in. | |
| 88.9 cm | |
| Premium protection for controlled environments and inherent barrier to dry particles, microorganisms, and non-hazardous liquids | |
| 104.14 cm | |
| 188.97 to 201.16 cm | |
| Class 1000 (ISO 6) |
| 3X-Large | |
| Tyvek | |
| Single-use | |
| Elastic | |
| Bound | |
| 46 to 52 in. | |
| 116.8 to 132.08 cm | |
| 5.4 mil | |
| 41 in. | |
| 6.2 to 6.6 ft. | |
| Clean-processed |