Learn More
Keystone Cleanroom Products DuPont 1073B Tyvek Autoclave Continuous Form Tubing
Superior seal strength for stoppers, seals and heavy equipment parts and minimal drying time. Simply cut Tyvek tubing to desired length and heat seal both ends closed using a validate-able impulse, or continuous band sealer.
Supplier:  Keystone Cleanroom Products TUBMGTVK28
 View more versions of this product
Catalog No. 19990176
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Medical grade 1073B Tyvek™ Tubing is breathable while providing an excellent bacterial, and microbial barrier for superior performance post steam sterilization
  • Lint free (Helmke Drum), no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 requirements
  • Heat sealed and packaged in an ISO 6 controlled clean room.
  • Designed for steam sterilization
  • Recommended for use with a heat sealer
  • Hydrophobic
  • Resists tearing and punctures
Specifications
Chemical Identifiers
Autoclavable
White
Steam Sterilization
Tyvek
Yes
<250°F
500 ft.
152 m
COC
Single-use
Pharmaceutical Equipment
Non-sterile
Steam
<125°C
Sterilization Reel
Specifications
Autoclavable
Heat Seal
Single-use
Pharmaceutical Equipment
Non-sterile
Steam
<125°C
500 ft.
Sterilization Reel
71 cm
COC
White
Steam Sterilization
Tyvek
Yes
<250°F
500 ft. x 28 in.
28 in.
152 m
Product Suggestions
Product Suggestions