Learn More
Fine Science Tools Dumont AA - Epoxy Coated Forceps, 12.5 cm length
Catalog No. NC9839858
Encompass
Slip-proof acid and electric shock resistant epoxy coating covers the forceps 17.5mm from the tips.
Catalog No. NC9839858 Supplier Fine Science Tools Supplier No. 1121010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Slip-proof acid and electric shock resistant epoxy coating covers the forceps 17.5mm from the tips.