Learn More
Fine Science Tools DUMONT FORCEPTS NO7 11.5CM
Catalog No. NC9484869
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumoxel/ Length: 11.5cm
Catalog No. NC9484869 Supplier Fine Science Tools Supplier No. 1127130
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumoxel/ Length: 11.5cm