missing translation for 'onlineSavingsMsg'
Learn More

Duloxetine Hydrochloride 98.0+%, TCI America™

Catalog No. D42231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42231G 1 g
D42235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42231G Supplier TCI America Supplier No. D42231G
Only null left

Chemical Identifiers

CAS 136434-34-9
Molecular Formula C18H20ClNOS
Molecular Weight (g/mol) 333.87
MDL Number MFCD06407958,MFCD06801358
InChI Key BFFSMCNJSOPUAY-LMOVPXPDSA-N
Synonym (S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride
PubChem CID 87834970
IUPAC Name hydrogen methyl[(3S)-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amine chloride
SMILES [H+].[Cl-].CNCC[C@H](OC1=C2C=CC=CC2=CC=C1)C1=CC=CS1

Specifications

Melting Point 164°C
Quantity 1 g
Formula Weight 333.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Duloxetine Hydrochloride
RTECSNumber XN0258000
TSCA No