missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Duke Standards™ 2000 Series Uniform Polymer Particles

Analyze wide size range materials through laser diffraction or other methods with uniform polymer particles that mimic real world particles.

Supplier:  Thermo Scientific™ 2007A

 View more versions of this product

Catalog No. 09980002

Only null left

Description

Description

Analyze wide size range materials through laser diffraction or other methods with Thermo Scientific™ Duke Standards™ 2000 Series Uniform Polymer Particles, NIST™ traceable polystyrene DVB particles with wider size distributions that mimic real world particles and work better on laser diffraction-type instruments. Particles meet the need for NIST traceable size standards that have slightly wider distributions than our monodisperse 3000/4000 series size standards. The wider distribution provides light scatter across a range of detectors, resulting in a more repeatable measurement.

Features:
  • Material is composed of polystyrene cross-linked with divinylbenzene, giving the particle good durability and chemical stability
  • Density of the polymer is 1.05g/cm3 and the index of refraction is 1.59 at 589nm (25°C)
  • Particles are packaged as an aqueous suspension in 15mL dropper-tipped vials
  • Excellent for use with any application that requires a NIST traceable size standard with a very narrow standard deviation
Specifications

Chemical Identifiers

Contains trace amount of surfactants
White or Colorless
Polystyrene Divinylbenzene (PS-DVB)
Material Safety Data Sheet (MSDS), Certificate of Calibration and Traceability to NIST
15 mL
=24 months
Store at room temperature or keep refrigerated. Keep bottle tightly sealed to avoid contamination.
Narrow Size Standard Deviation Applications
Polymer Particles
1.05g/cm³
1.59 at 589nm (25°C)

Specifications

Contains trace amount of surfactants
White or Colorless
Polystyrene Divinylbenzene (PS-DVB)
Material Safety Data Sheet (MSDS), Certificate of Calibration and Traceability to NIST
15 mL
s = 0.4 µm, 5.7% CV
7 μm
Store at room temperature or keep refrigerated. Keep bottle tightly sealed to avoid contamination.
Narrow Size Standard Deviation Applications
Polymer Particles
1.05g/cm³
1.59 at 589nm (25°C)
=24 months
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions