Learn More
LaMotteā„¢ Dual Range QAC Test Strips
Dual Range QAC Test Strip takes care of all testing needs on one strip.
Supplier:  LaMotteā„¢ 2934
Catalog No. S43599
Only null left
Explore available promotions
Description
Description
Whether checking the concentration of a quaternary ammonium compound solution or checking to ensure complete removal, the Dual Range QAC test strip is ideal. Chemically treated test strips change color to indicate sanitizer level. Strips and color chart are packaged in a waterproof plastic vial.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Yes
50 Tests
Test Strip
Food and beverage, laundry and sanitation, industrial water boiling and cooling, water and wastewater
Low range: 0, 10, 20, 40, 80; High range: 0, 100, 200, 400, 800
Product Suggestions
Product Suggestions