Learn More
Hoefer Inc Running Tray w/Silicone Gasket for SUB15
Catalog No. NC2957552
Encompass
SE6015 Dual Gel Caster is included when you order an SE600 Series unit.It can also be ordered separately.Casts two 8, 16, or 24 cm long gels at a time;Double the capacity with optional divider plates;Easy to use and leak free;Glass sandwiches seal into the casting cradle with a simple twist of the cams;Includes: Caster Stand with 2 Cradles and Leveling Feet; Laminated Gaskets2 pcs
Catalog No. NC2957552 Supplier Hoefer Inc Supplier No. SE6015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SE6015 Dual Gel Caster is included when you order an SE600 Series unit.It can also be ordered separately.Casts two 8, 16, or 24 cm long gels at a time;Double the capacity with optional divider plates;Easy to use and leak free;Glass sandwiches seal into the casting cradle with a simple twist of the cams;Includes: Caster Stand with 2 Cradles and Leveling Feet; Laminated Gaskets2 pcs