missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ Dual E-cylinder Manifold

Catalog No. 04777124
Change view
Click to view available options
Length (Metric):
26.7 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
04777124 26.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777124 Supplier SOMNI Scientific™ Supplier No. RE20100

Can be used with a single or two E size oxygen cylinders

  • The incorporated check valves prevent leakage when one cylinder is disconnected
  • Preset regulator, mounting hardware, oxygen hose and cylinder wrench included
  • Attach one tank or two
  • Check valves incorporated
  • Preset regulator delivers 50-55 psi
  • Durable and easy to install

Specifications

For Use With (Application) Animal Oxygen Supply, Inhalant Anesthesia
For Use With (Equipment) RM-17003, RM-17004, OC-21003, OC-21004, MW-17005
Material Stainless Steel, Aluminum
Length (Metric) 26.7 cm
Width (Metric) 15.2 cm
Includes Preset regulator, mounting hardware, oxygen hose and cylinder wrench

For Veterinary Use Only