missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ Dual Diverter Valve System

Catalog No. 04777110
Encompass
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04777110 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777110 Supplier SOMNI Scientific™ Supplier No. DS14006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Color coded inhalant anesthesia delivery adaptable to most standard anesthesia systems.

  • A cost-effective way to add an additional anesthesia delivery line to your current anesthesia system
  • Adaptable to most anesthesia systems
  • Converts one delivery line to two
  • Cost effective
  • Color coded, adapters included

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) TT-17000, TT-17001, TT-17002
Material Plastic
Length (Metric) 213.36 cm
Width (Metric) 0.635 cm
Includes Dual diverters with adapters

For Veterinary Use Only