Learn More
Mopec Dual Approach Wall Mount Autopsy Sink
Save time and space with wall mounted autopsy dissection sink. Mopec™ Dual Approach Wall Mount Autopsy Sink features a cart tray that hangs over the sink to allow for proper fluid drainage.
Supplier:  Mopec CC300
Catalog No. 22444017
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Stainless steel construction
  • Hydro aspirator with reversing flow valve
  • Includes hose hanger and 8 foot clear PVC tubing
  • Two cold water body rinse valve with vacuum breaker and serrated hose fitting
  • Two hot and cold water fixtures with wrist blade handles
  • 8 in. swing spout with vacuum breaker
  • Three interchangeable removable stainless steel grid plates
  • Two GFCI duplex outlets with waterproof covers
  • Two instrument drawers- Flush to the table when closed and fitted
  • Flush inset handles at the top to keep fluid from accumulating in the drawers
  • Two hand spray assemblies
Warranty
1 Year Warranty
Specifications
Specifications
Autopsy and Embalming Sink
38 in.
71 cm
150 in.
154 cm
96 cm
381 cm
Stainless Steel
6 in.
15 cm
61 in.
38 in.
20 in.
50 cm
Product Suggestions
Product Suggestions