missing translation for 'onlineSavingsMsg'
Learn More

DTT, Molecular Grade, Promega™

Catalog No. PRV3155
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3155 25 g
PRV3151 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3155 Supplier Promega Corporation Supplier No. V3155

An antioxidant used to stabilize enzymes and other proteins containing sulfhydryl groups.

  • Quality tested and certified free of DNase, RNase and protease activity
  • Available as a liquid (100μL) or as a powder (5g or 25g)
  • Supplied at a concentration of 100mM

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Color White
Quantity 25 g
Formula Weight 154.25
Grade Molecular
Physical Form Crystals, Powder or Liquid
Chemical Name or Material DTT (Dry Powder)
Recommended Storage Store -20°C.