missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTT, Molecular Grade, Promega™
Description
- Quality tested and certified free of DNase, RNase and protease activity
- Available as a liquid (100μL) or as a powder (5g or 25g)
- Supplied at a concentration of 100mM
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 154.25 |
| Grade | Molecular |
| Physical Form | Crystals, Powder or Liquid |
| Chemical Name or Material | DTT (Dry Powder) |
Safety and Handling
| Recommended Storage | Store -20°C. |