Research Products Corporation DTT [DL-Dithiothreitol] (Cleland's Reagent)], 5 Grams
Catalog No. 50213272
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.
Catalog No.
50213272
Supplier
Research Products CorporationSupplier No.
RPI D110005.0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.