Learn More
Synthware™ Drying Tube, 90° Bent, Microscale
GREENER_CHOICE
Catalog No. 31502101
Change view
Click to view available options
Standard Taper Joint No.:
14/10
14/20
19/22
24/40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31502101 14/10
31500646 14/20
31500647 19/22
31500648 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31502101 Supplier Synthware™ Supplier No. A851410
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Drying tube is used to protect moisture sensitive reactions from atmosphere while permitting system to remain unsealed. For microscale applications
  • For protection of moisture sensitive reactions from atmosphere
  • Joint 14/10
Specifications
Material Borosilicate Glass (3.3)
Product Type 90° Bent Drying tube
For Use With (Application) Organic Synthesis
Standard Taper Joint No. 14/10