Learn More
Synthware™ Drying Tube, 90° Bent, Microscale
Drying tube is used to protect moisture sensitive reactions from atmosphere while permitting system to remain unsealed. For microscale applications
Supplier: Synthware™ A852440
Description
- For protection of moisture sensitive reactions from atmosphere
- Joint 14/10
Chemical Identifiers
| 90° Bent Drying tube | |
| Organic Synthesis |
Specifications
| Organic Synthesis | |
| 90° Bent Drying tube | |
| Borosilicate Glass | |
| 24/40 |