Learn More
Synthware™ Drying Tube, 90° Bent, Microscale
Description
- For protection of moisture sensitive reactions from atmosphere
- Joint 14/10
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Product Type | 90° Bent Drying tube |
| Material | Borosilicate Glass |
| Standard Taper Joint No. | 19/22 |