missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Drying Pearls Orange

Catalog No. 94098500G
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94098500G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 94098500G Supplier Sigma Aldrich Supplier No. 94098500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

from orange to colorless after moisture absorption

Specifications

CAS 79917-90-1
Linear Formula C8H15ClN2
MDL Number MFCD03098976
Quantity 1 kg
Synonym Aluminum silicate; Silica gel orange; Aluminosilicate