Learn More
Anton Paar Drying Cartridge for Internal Air Pump
Catalog No. NC0071442
Encompass
Drying cartridge for absorbing humidity from ambient air. Use to prevent condensation if drying with the air pump.Suitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230
Catalog No. NC0071442 Supplier Anton Paar Supplier No. ANP 65085
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Drying cartridge for absorbing humidity from ambient air. Use to prevent condensation if drying with the air pump.Suitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230