Learn More
Beckman Coulter Dry Powder System (DPS) HiCapacity HEPA Vacuum Unit and Accessories
The HiCap HEPA Vacuum Unit is designed for use with the Dry Powder System (DPS) of the LS 13 320 XR Particle Size Analyzer. The Vacuum Unit delivers dry powder samples to the sensing zone in the optical bench of the LS 13 320 XR, all required connectors are included.
Supplier:  Beckman Coulter C06826
 View more versions of this product
Catalog No. 15655169
Only null left
Description
Description
  • LS 13 320 XR Particle Size Analyzers equipped with the Dry Powder System (DPS) Module require a vacuum unit for use
  • HiCap HEPA Vacuum Units deliver dry powder samples to the sensing zone in the optical bench
  • Vacuums feature a fully adjustable pressure range
  • Replacement HiCap Vacuum Filters are available
Specifications
Chemical Identifiers
LS 13 320 XR and Dry Powder System (DPS)
Specifications
HiCapacity HEPA Vacuum Unit
Delivering Dry Powder Samples to the Sensing Zone in the Optical Bench of LS 13 320 XR
LS 13 320 XR and Dry Powder System (DPS)
Product Suggestions
Product Suggestions