Learn More
Drummond™ Dispensing Manifold
Catalog No. 2116911A
Change view
Click to view available options
Angle:
No
Yes
Number of Channels:
12
8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Angle Number of Channels
2116911A No 12
2116910D No 8
2116910E Yes 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2116911A Supplier Drummond™ Supplier No. 3000096
Only null left
Eight-tip manifolds for use with the 21-169-10A Digital Microdispenser or any pipetter that accepts standard Luer-Lok™ connector
  • Stainless steel and PTFE construction
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Specifications
Angle No
Description 9mm (center to center); 11mm pin length
Adjustable Tip Spacing 9 mm to 11 mm
For Use With Drummond MicroPipette
Material Stainless Steel, PTFE
Number of Channels 12
For Use With (Equipment) Drummond™ micropipetter
Includes Cleaning wire, Luer connector and end caps