missing translation for 'onlineSavingsMsg'
Learn More

Drummond™ Portable Pipet-Aid Pipette Controller Accessory, Charger

Catalog No. 1368117e
Change view
Click to view available options
:
Charger, 110V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
1368117E Charger, 110V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1368117E Supplier Drummond™ Supplier No. 4000030
Only null left

Ensure quality and performance by selecting the manufacturer's component parts. Drummond Pipet-Aid Charger works with the Portable Pipet-Aid series of pipette controllers.

  • 110V (8.7V/8mA)

Specifications

Autoclavable Not Autoclavable
Type Charger
For Use With (Equipment) Original Portable Pipet-Aid, Portable Pipet-Aid XP2
Voltage 110 V
Amperage 8mA