Learn More
Drummond™ Pipet-Aid™ Series Pipet Controller Accessory, Ni-MH Rechargeable Battery
For use with Pipet-Aid™ Series Pipet Controller
Supplier: Drummond™ 4000035
Description
Ni-MH Rechargeable Battery for use with Pipet-Aid™ Series Pipet Controller
Specifications
| NiMH | |
| Original Portable Pipet-Aid, and Portable Pipet-Aid XP | |
| 8.4 Volt |
| Not Autoclavable | |
| 1 Each |