Learn More
Drummond™ PCR Micropipets
Disposable, positive displacement micropipets with stainless steel plungers
$235.28 - $235.28
Specifications
| For Use With (Application) | To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols |
|---|---|
| Type | PCR Micropipet |
| Disposable | Single-use |
Description
- Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
- Decontaminate at temperature of 400°F or higher
Specifications
| To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols | |
| PCR Micropipet | |
| Single-use |