Learn More
Drummond™ PCR Micropipets
Catalog No. 21179B
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
Catalog No. Volume (Metric)
21179B 1 to 10 μL
1 options
Catalog No. 21179B Supplier Drummond™ Supplier No. 50001001X10
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable, positive displacement micropipets with stainless steel plungers
  • Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
  • Decontaminate at temperature of 400°F or higher
  • Stainless steel plungers
  • To deliver 1-10μL
Specifications
For Use With (Application) To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
Includes 100 disposable bores, 100 disposable plungers
Quantity 100 Micropipets
Volume (Metric) 1 to 10 μL
Type PCR Micropipet
Disposable Single-use