Learn More
Drummond™ Original Portable Pipet-Aid™ Pipet Controller
Catalog No. 1368119
Change view
Click to view available options
Voltage:
110 V
Catalog No. Voltage
1368119 110 V
1 options
Catalog No. 1368119 Supplier Drummond™ Supplier No. 4000100
Only null left
Convenient alternative to mouth pipetting in the laboratory.
  • Easy-to-use volume control mechanism permits precise fluid intake and delivery
  • Check valve protects against accidental fluid intake into the handle
  • Can be used with glass or plastic pipets
  • Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
Specifications
Autoclavable Autoclavable
Type Portable Filler/Dispenser
For Use With (Application) Laboratory pipetting
For Use With (Equipment) Glass or plastic pipets
Includes Pipet Controller, TC Nosepiece
Voltage 110 V