Learn More
Drummond™ Fixed Volume Microdispensers Replacement Parts Kits
Supplier: Drummond™ 3000005K
Description
Replacement Parts Kits for Microdispensers. For contents of each kit, see chart below.
Specifications
| 5 μL | |
| Drummond™ Fixed Volume Microdispenser | |
| 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide |