missing translation for 'onlineSavingsMsg'
Learn More

Drummond™ Calibrated Micropipets

Anticoagulant treatment for clinical applications

Supplier:  Drummond™ 2000020

 View more versions of this product

Catalog No. 21-180-14

Only null left
Explore available promotions

Description

Description

Anticoagulant treatment for clinical applications

  • Ideal for liquid transfer work
  • Fire polished
  • Accuracy ±0.5%
  • Color coded
  • Use heparinized 44.7μL pipet for blood transfer work
  • Available in sizes from 1 to 200μL

Order Info

Supplied in pack of 250 micropipets

Specifications

Specifications

±0.5%
Glass
250 pipets and one 15 in. aspirator tube
20 μL
Yes
Anticoagulant treatment for clinical applications, ideal for liquid transfer applications
Black
Calibrated Micropipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions