missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Drum Storage Cabinets
GREENER_CHOICE

Catalog No. 18887C
Change view
Click to view available options
Dimensions (D x W x H) Interior:
28.25 x 28.25 x 62 in.
28.5 x 55 x 62 in.
Width (English) Exterior:
31.25 in.
58 in.
Weight (English):
334 lb.
575 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior Width (English) Exterior Weight (English)
18887C 28.5 x 55 x 62 in. 58 in. 575 lb.
171561A 28.25 x 28.25 x 62 in. 31.25 in. 334 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18887C Supplier Eagle™ Supplier No. HAZ1955

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Drum supports and built-in grounding connectors. Cabinets for hazardous waste drums include drum rollers for easy drum handling.

The Eagle Drum Storage Cabinet is designed to let you store 55-gallon drums containing hazardous waste or flammables.

  • Safely store hazardous waste or flammables in the workplace
  • One- and two-drum capacity with choice of manual or self-closing doors
  • Store drums vertically or horizontally
  • Vertical cabinets include half shelf for additional storage of accessories or containers
  • Cabinets feature large red FLAMMABLE - KEEP FIRE AWAY and HAZARDOUS WASTE labels on yellow enamel finish; flammables cabinets have FLAMMABLE - KEEP FIRE AWAY label

Certifications

FM approved (except 17-516-1B).

Compliance

Comply with NFPA Code 30, and meet OSHA 29CFR1910.106(d)3 standard.

‡Accommodates no more than 60 gal. (227L) may be Class I and Class II liquids.

†Not FM approved.

Specifications

Dimensions (D x W x H) Interior 28.5 x 55 x 62 in.
Product Type Vertical drum storage cabinet
Description Haz-Mat, two drum, two door, manual close doors
Height (Metric) Exterior 165.1 cm
Depth (Metric) Exterior 79.37 cm
Weight (Metric) 260.08 kg
Depth (Metric) Interior 72.39 cm
Width (Metric) Interior 139.7 cm
No. of Doors 2
Width (Metric) Exterior 147 cm
No. of Shelves 1
Height (Metric) Interior 157.4 cm
Load Bearing Capacity 110 gal.
Material Steel
Width (English) Exterior 58 in.
Width (English) Interior 55 in.
Certifications/Compliance FM, NFPA, OSHA
Height (English) Exterior 65 in.
Door Style Manual-close
Color Yellow
Includes Drum supports and built-in grounding connectors, drum rollers, one shelf, hazardous waste labels
Height (English) Interior 62 in.
For Use With (Application) Flammables
Depth (English) Interior 28.5 in.
Weight (English) 575 lb.
Depth (English) Exterior 31.25 in.
Show More Show Less