missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 55 gal., Self-Close, with Drum Support

Catalog No. p-200157454
Change view
Click to view available options
Color:
Blue
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18666741 Blue
18666742 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18666741 Supplier JUSTRITE Supplier No. 896222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Width (English) Interior 30.8 in.
Width (Metric) Interior 864 mm
Weight (Metric) 157.4 kg
No. of Doors 2
Surface Coating Powder-coat Paint
Width (Metric) Exterior 781 mm
No. of Shelves 1
Weight (English) 347 lb.
Load Bearing Capacity 350 lb./shelf
Height (Metric) Exterior 1651 mm
Height (Metric) Interior 1537 mm
Explosion Relief Panel No
Height (English) Interior 60.5 in.
Product Type Vertical Drum Storage Cabinet
Door Style Self-close, Locking, Three-point lock
Width (English) Exterior 34 in.
Color Blue
Height (English) Exterior 65 in.
Material Steel (18 ga), Galvanized Steel
Dimensions (D x W x H) Interior 30.3 x 30.8 x 60.5 in.
Dimensions (D x W x H) 34 x 34 x 65 in.
Depth (English) Exterior 34 in.
Depth (Metric) Interior 771 mm
Certifications/Compliance FM Approved
Depth (English) Interior 30.8 in.
Depth (Metric) Exterior 864 mm
Show More Show Less