missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 55 gal., Self-Close, with Drum Rollers

Catalog No. p-200157453
Change view
Click to view available options
Color:
Blue
Gray
Green
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18666751 Blue
18666752 Gray
18666753 Green
18666754 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18666751 Supplier JUSTRITE Supplier No. 896272

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Surface Coating Powder-coat Paint
No. of Shelves 1
Weight (Metric) 161 kg
Weight (English) 355 lb.
Width (English) Interior 30.8 in.
Width (Metric) Exterior 864 mm
Width (Metric) Interior 781 mm
No. of Doors 2
Door Style Self-close, Locking, Three-point lock
Explosion Relief Panel No
Load Bearing Capacity 350 lb./shelf
Product Type Vertical Drum Storage Cabinet
Material Steel (18 ga), Galvanized Steel
Width (English) Exterior 34 in.
Color Blue
Height (English) Exterior 65 in.
Certifications/Compliance FM Approved
Depth (English) Exterior 34 in.
Show More Show Less