missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JUSTRITE Drum Storage Cabinets, 55 gal., Manual Close, with Drum Support
Description
Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.
- 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
- Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
- Available in manual or self-close models.
- Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
- Independently fire tested and approved by FM Approvals.
- High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
- U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
- Haz-Alert™ reflective warning labels are highly visible under fire conditions.
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
- Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty
Specifications
Specifications
| Width (Metric) Exterior | 781 mm |
| Width (Metric) Interior | 864 mm |
| Weight (English) | 359 lb. |
| Load Bearing Capacity | 350 lb./shelf |
| Surface Coating | Powder-coat Paint |
| Height (Metric) Interior | 1537 mm |
| Weight (Metric) | 162.8 kg |
| No. of Shelves | 1 |
| No. of Doors | 2 |
| Width (English) Interior | 30.8 in. |
| Show More |