missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 55 gal., Manual Close, with Drum Support

Catalog No. 18666735
Change view
Click to view available options
Color:
Gray
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18666735 Red
18666737 Gray
18666738 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18666735 Supplier JUSTRITE Supplier No. 896201

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Height (English) Interior 60.5 in.
Width (Metric) Exterior 781 mm
No. of Shelves 1
Width (English) Interior 30.8 in.
Width (Metric) Interior 864 mm
Weight (English) 359 lb.
Height (Metric) Exterior 1651 mm
Load Bearing Capacity 350 lb./shelf
Weight (Metric) 162.8 kg
Surface Coating Powder-coat Paint
Height (Metric) Interior 1537 mm
No. of Doors 2
Explosion Relief Panel Yes
Width (English) Exterior 34 in.
Door Style Manual-close, Locking, Three-point lock
Material Steel (18 ga), Galvanized Steel
Product Type Vertical Drum Storage Cabinet
Dimensions (D x W x H) Interior 30.3 x 30.8 x 60.5 in.
Height (English) Exterior 65 in.
Dimensions (D x W x H) 34 x 34 x 65 in.
Depth (English) Interior 30.3 in.
Certifications/Compliance FM Approved
Depth (Metric) Interior 771 mm
Color Red
Depth (English) Exterior 34 in.
Depth (Metric) Exterior 864 mm
Show More Show Less