missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 100 gal., Manual Close, with Drum Support

Catalog No. p-200157449
Change view
Click to view available options
Color:
Blue
Gray
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18666755 Red
18666756 Blue
18666757 Gray
18666758 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18666755 Supplier JUSTRITE Supplier No. 899101*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Surface Coating Powder-coat Paint
Weight (English) 555 lb.
Width (Metric) Exterior 1499 mm
Width (Metric) Interior 1422 mm
Width (English) Interior 56 in.
Weight (Metric) 249.5 kg
No. of Shelves 1
Width (English) Exterior 59 in.
Material Steel (18 ga), Galvanized Steel
Color Red
Product Type Vertical Drum Storage Cabinet
Depth (English) Exterior 59 in.
Height (English) Exterior 65 in.
Show More Show Less