missing translation for 'onlineSavingsMsg'
Learn More

Droxidopa 98.0+%, TCI America™

Catalog No. d4235200mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
200 mg
50 mg
Catalog No. Quantity
D4235200MG 200 mg
D423550MG 50 mg
2 options

Catalog No. D4235200MG

Supplier: TCI America D4235200MG

Only null left

Chemical Identifiers

CAS 23651-95-8
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00799030
InChI Key QXWYKJLNLSIPIN-JGVFFNPUSA-N
Synonym (2S,3R)-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid, L-threo-3,4-Dihydroxyphenylserine
PubChem CID 92974
ChEBI CHEBI:31524
IUPAC Name (2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid
SMILES C1=CC(=C(C=C1C(C(C(=O)O)N)O)O)O

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 200 mg
Formula Weight 213.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Droxidopa
RTECSNumber VT9626010
TSCA No