missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Droxidopa 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 225°C |
| Color | White-Yellow |
| Quantity | 50 mg |
| Formula Weight | 213.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Droxidopa |
Safety and Handling
| RTECSNumber | VT9626010 |
| TSCA | No |