Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
 View more versions of this product
Catalog No. AS275
Only null left
Description
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
Chemical Identifiers
Cellulose Acetate
Minimizes evaporation, keeps food fresh longer
1000 Case
BuzzPlugs Bottle Closure
Superior barrier
Specifications
BuzzPlugs Bottle Closure
Superior barrier
Minimizes evaporation, keeps food fresh longer
Cellulose Acetate
2.8cm wide plastic shell vials
1000 Case
Product Suggestions
Product Suggestions