Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Catalog No. AS273
Change view
Click to view available options
For Use With (Equipment):
2.5cm narrow plastic shell vials
2.8cm wide plastic shell vials
Standard glass vials
Standard plastic stock bottles
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
AS273 2.5cm narrow plastic shell vials
AS275 2.8cm wide plastic shell vials
AS271 Standard glass vials
AS277 Standard plastic stock bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AS273
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
Type BuzzPlugs Bottle Closure
For Use With (Application) Minimizes evaporation, keeps food fresh longer
For Use With (Equipment) 2.5cm narrow plastic shell vials
Quantity 1000 Case
Material Cellulose Acetate
Includes Superior barrier